Learn More
Aviva Systems Biology Corp BMP2K Antibody - C-terminal region (ARP42483_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42483_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BMP2K. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human BMP2K. Alternate names: BIKE, DKFZp434K0614, DKFZp434P0116, HRIHFB2017ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58242