Learn More
Aviva Systems Biology Corp BMP2K Antibody - middle region (ARP42482_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42482_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BMP2K. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BMP2K. Alternate names: BIKE, DKFZp434K0614, DKFZp434P0116, HRIHFB2017ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-58241