Learn More
Aviva Systems Biology Corp BMP4 Antibody - N-terminal region (ARP63063_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63063_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BMP4. It was validated on Western Blot. . Alternate names: BMP2B, BMP2B1, MCOPS6, OFC11, ZYMEApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-287-860