Learn More
Aviva Systems Biology Corp BMP6 Antibody - N-terminal region (ARP58756_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58756_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BMP6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BMP6. Alternate names: VGR, VGR1ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-286-495