Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ BMP7 (Human) ELISA Kit

Catalog No. 89100729 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Bone Tissue, CCS, Plasma, Serum, Urine
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-729 Bone Tissue, CCS, Plasma, Serum, Urine
89-028-435 CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-729 Supplier Abnova™ Supplier No. KA3022
Add to Cart
Add to Cart

In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human BMP7

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description BMP7 (Human) ELISA Kit is intended for the quantitative measurement of human BMP7.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human BMP-7 antibody, Lyophilized BMP-7 standards (10ng x 2), Standard diluent buffer (30mL), Biotin conjugated anti-Human Activin A antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), 25X Wash buffer (30mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Bone Tissue, CCS, Plasma, Serum, Urine
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type BMP7 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.