Learn More
Aviva Systems Biology Corp BMP8A Antibody - N-terminal region (ARP65414_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65414_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BMP8A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human BMP8A. Alternate names: FLJ14351, FLJ45264ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-494