Learn More
Aviva Systems Biology Corp BMPER Antibody - C-terminal region (ARP52569_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52569_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BMPER. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human BMPER. Alternate names: CRIM3, CV-2, CV2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53698