Learn More
Carnabio Usa Inc BMPR1A/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 091375
Encompass
BMPR1A, Catalytic domain, Active mutant [Q223D], Amino Acid 187-532, NP_004320.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0340