Learn More
Carnabio Usa Inc BMPR2/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 091395
Encompass
BMPR2, Cytoplasmic domain, Wild type, Amino Acid 174-1038, NP_001195.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0342