Learn More
New England Biolabs, Inc. BmrI – 100 units
SDP
Catalog No. 50812322
Encompass
A restriction endonuclease that recognizes the sequence ACTGGGNNNN_N^.
Catalog No. 50-812-322 Supplier New England Biolabs, Inc. Supplier No. R0600S
A restriction endonuclease that recognizes the sequence ACTGGGNNNN_N^.