Learn More
Being Scientific BMS-07B3 - 3L B, 5.1×5.1(inch), Ambient +5℃ to 320℃, 200 - 2000rpm
Catalog No. 5000217561
Encompass_Preferred
The BEING Magnetic Heated Stirrers offer a large variety of sizes and temperature ranges to accommodate your laboratory needs. Our stirrers provide a user friendly PID controller offering a large LCD color screen for easy viewing. They are constructed for durability, high performance and safety.
Catalog No. 50-002-17561 Supplier Being Scientific Supplier No. BMB150703U
The BEING Magnetic Heated Stirrers offer a large variety of sizes and temperature ranges to accommodate your laboratory needs. Our stirrers provide a user friendly PID controller offering a large LCD color screen for easy viewing. They are constructed for durability, high performance and safety.