Learn More
Being Scientific BMS-09B15 - 15L, 7.1 x 7.1 inch, Ambient +5℃ to 320℃, 200 - 2000rpm
Catalog No. 5000217578
Encompass_Preferred
The BEING Magnetic Heated Stirrers offer a large variety of sizes and temperature ranges to accommodate your laboratory needs. Our stirrers provide a user friendly PID controller offering a large LCD color screen for easy viewing. They are constructed for durability, high performance and safety.
Catalog No. 50-002-17578 Supplier Being Scientific Supplier No. BMB150915U
The BEING Magnetic Heated Stirrers offer a large variety of sizes and temperature ranges to accommodate your laboratory needs. Our stirrers provide a user friendly PID controller offering a large LCD color screen for easy viewing. They are constructed for durability, high performance and safety.