Learn More
Selleck Chemical LLC BMS-1-5mg
SDP
Catalog No. 502390061
Encompass_Preferred
BMS-1 is a small-molecule inhibitor of PD-1/PD-L1 interaction with IC50 of 6 nM.
Catalog No. 50-239-0061 Supplier Selleck Chemical LLC Supplier No. S79115MG
Add to Cart
Add to Cart
BMS-1 is a small-molecule inhibitor of PD-1/PD-L1 interaction with IC50 of 6 nM.