Learn More
Medchemexpress LLC HY-120635 5mg Medchemexpress, BMS-1001 (hydrochloride) CAS:2113650-04-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1206355MG
Encompass_Preferred
Medchemexpress, HY-120635 5mg BMS-1001 (HCl) CAS:2113650-04-5 BMS-1001 hydrochloride is an orally active human PD-L1/PD-1 immune checkpoint inhibitor. BMS-1001 hydrochloride exhibits low-toxicity in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9610