Learn More
Medchemexpress LLC BMS-1001 hydrochloride | 2113650-04-5 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12063510MG
Encompass_Preferred
BMS-1001 hydrochloride is a small-molecule PD-L1/PD-1 immune checkpoint inhibitor supplied for research use. It binds human PD-L1 and blocks PD-1/PD-L1 interaction, and is used in immunology and cancer research assays.
  • Small-molecule PD-L1/PD-1 inhibitor for research use.
  • Reported IC50 of 2.25 nM in HTRF binding assay.
  • Available as solid and solution forms; common solid sizes include 5 mg-100 mg.
  • Typical purity approximately 98.0% by supplier assay.
  • Molecular weight 631.11 g/mol and formula C35H35ClN2O7.
Catalog No. 50-002-32068