Learn More
Medchemexpress LLC BMS-1166 (hydrochloride) | 2113650-05-6 | 100.0% | 677.57 g/mol | C36H34Cl2N2O7 | 5 MG
SDP
Catalog No. 5000246716
Encompass_Preferred
BMS-1166 hydrochloride is a small-molecule PD-1/PD-L1 immune checkpoint inhibitor that induces PD-L1 dimerization and blocks PD-1 binding. It exhibits potent biochemical activity in the low-nanomolar range and is supplied as a stable hydrochloride salt in solid form for research use in biochemical and cellular assays.
  • Potent PD-1/PD-L1 interaction inhibitor with low-nanomolar activity.
  • Hydrochloride salt, solid, white to off-white, suitable for handling.
  • High purity appropriate for research applications.
  • Stable as a powder under recommended storage conditions for extended shelf life.
  • Intended for use in biochemical and cell-based immune checkpoint studies.
Catalog No. 50-002-46716 Supplier Medchemexpress LLC Supplier No. HY102011A5MG
BMS-1166 hydrochloride is a small-molecule PD-1/PD-L1 immune checkpoint inhibitor that induces PD-L1 dimerization and blocks PD-1 binding. It exhibits potent biochemical activity in the low-nanomolar range and is supplied as a stable hydrochloride salt in solid form for research use in biochemical and cellular assays.
  • Potent PD-1/PD-L1 interaction inhibitor with low-nanomolar activity.
  • Hydrochloride salt, solid, white to off-white, suitable for handling.
  • High purity appropriate for research applications.
  • Stable as a powder under recommended storage conditions for extended shelf life.
  • Intended for use in biochemical and cell-based immune checkpoint studies.