Learn More
Medchemexpress LLC HY-108593 10mg Medchemexpress, BMS-191011 CAS:202821-81-6 Purity:>98%
SDP
Catalog No. 501968586
Encompass_Preferred
Medchemexpress, HY-108593 10mg BMS-191011 CAS:202821-81-6 BMS-191011 (BMS-A) is an opener of the large-conductance, Ca2++-activated potassium (maxi-K) channel, effective in stroke models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8586 Supplier Medchemexpress LLC Supplier No. HY10859310MG
Medchemexpress, HY-108593 10mg BMS-191011 CAS:202821-81-6 BMS-191011 (BMS-A) is an opener of the large-conductance, Ca2++-activated potassium (maxi-K) channel, effective in stroke models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.