Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-192364 50mg | 202822-21-7 | 355.70 g·mol⁻¹ | C15H9ClF3N3O2 | 50 MG
SDP

Catalog No. 5000226833
Encompass_Preferred

BMS-192364 is a small-molecule research compound described as an RGS (regulator of G-protein signaling) agonist. It modulates Gα-RGS interactions and alters calcium flux, and has been used in studies of urinary bladder contraction and G-protein-coupled signaling pathways. Supplied as a high-purity 50 mg vial for research applications.

  • Targets Gα-RGS interaction as an RGS agonist.
  • Modulates calcium signaling and reduces bladder contraction in models.
  • Molecular formula C15H9ClF3N3O2.
  • Molecular weight 355.70 g·mol⁻¹.
  • Supplied purity 99.01% and packaged as 50 mg.

Catalog No. 50-002-26833 Supplier Medchemexpress LLC Supplier No. HY11486550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-192364 is a small-molecule research compound described as an RGS (regulator of G-protein signaling) agonist. It modulates Gα-RGS interactions and alters calcium flux, and has been used in studies of urinary bladder contraction and G-protein-coupled signaling pathways. Supplied as a high-purity 50 mg vial for research applications.

  • Targets Gα-RGS interaction as an RGS agonist.
  • Modulates calcium signaling and reduces bladder contraction in models.
  • Molecular formula C15H9ClF3N3O2.
  • Molecular weight 355.70 g·mol⁻¹.
  • Supplied purity 99.01% and packaged as 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.