Learn More
Medchemexpress LLC BMS-193885 | 186185-03-5 | 99.9% | 5 MG
SDP
Catalog No. 5000234382
Encompass_Preferred
BMS-193885 is a research-grade selective neuropeptide Y1 receptor antagonist used in preclinical studies of appetite regulation and metabolic disease. It is blood-brain barrier permeable, supplied as solid or as DMSO solutions, and is commonly available in small research pack sizes such as 5 mg.
  • Selective neuropeptide Y1 receptor antagonist (Ki = 3.3 nM).
  • High purity (99.9% by HPLC).
  • Blood-brain barrier permeable, suitable for in vivo studies.
  • Available as solid and as pre-made DMSO solutions for stock preparation.
  • Common pack sizes include 5 mg for small-scale experiments.
Catalog No. 50-002-34382 Supplier Medchemexpress LLC Supplier No. HY1206195MG
BMS-193885 is a research-grade selective neuropeptide Y1 receptor antagonist used in preclinical studies of appetite regulation and metabolic disease. It is blood-brain barrier permeable, supplied as solid or as DMSO solutions, and is commonly available in small research pack sizes such as 5 mg.
  • Selective neuropeptide Y1 receptor antagonist (Ki = 3.3 nM).
  • High purity (99.9% by HPLC).
  • Blood-brain barrier permeable, suitable for in vivo studies.
  • Available as solid and as pre-made DMSO solutions for stock preparation.
  • Common pack sizes include 5 mg for small-scale experiments.