Learn More
Medchemexpress LLC BMS-199264 hydrochloride | 186180-83-6 | 99.3% | 567.53 g·mol⁻¹ | C26H32Cl2N4O4S | 100 MG
SDP
Catalog No. 5000308072
Encompass_Preferred
BMS-199264 hydrochloride is a research compound that selectively inhibits the F1F0 ATP hydrolase (reported IC50 = 0.5 μM), preventing ATP decline during ischemia and reducing cardiac necrosis while improving contractile recovery after reperfusion.
  • Inhibits F1F0 ATP hydrolase with reported IC50 of 0.5 μM.
  • Reduces ATP decline during ischemic conditions.
  • Helps limit cardiac necrosis and supports functional recovery.
  • Supplied as a hydrochloride salt with high reported purity.
  • Available in multiple pack sizes to suit experimental needs.
Catalog No. 50-003-08072 Supplier Medchemexpress LLC Supplier No. HY118960100MG
BMS-199264 hydrochloride is a research compound that selectively inhibits the F1F0 ATP hydrolase (reported IC50 = 0.5 μM), preventing ATP decline during ischemia and reducing cardiac necrosis while improving contractile recovery after reperfusion.
  • Inhibits F1F0 ATP hydrolase with reported IC50 of 0.5 μM.
  • Reduces ATP decline during ischemic conditions.
  • Helps limit cardiac necrosis and supports functional recovery.
  • Supplied as a hydrochloride salt with high reported purity.
  • Available in multiple pack sizes to suit experimental needs.