Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-199264 hydrochloride | 186180-83-6 | 99.3% | 567.53 g/mol | C26H32Cl2N4O4S | 25 MG
SDP

Catalog No. 5000228624
Encompass_Preferred

BMS-199264 hydrochloride is a research-grade small-molecule inhibitor of the F1F0 ATP hydrolase provided as a high-purity solid for biochemical and cardiac ischemia/reperfusion studies in vitro and in vivo.

  • High purity (99.28%).
  • Molecular weight 567.53 g/mol.
  • Chemical formula C26H32Cl2N4O4S.
  • Supplied as a 25 MG package.
  • Appearance: solid; storage: 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-28624 Supplier Medchemexpress LLC Supplier No. HY11896025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-199264 hydrochloride is a research-grade small-molecule inhibitor of the F1F0 ATP hydrolase provided as a high-purity solid for biochemical and cardiac ischemia/reperfusion studies in vitro and in vivo.

  • High purity (99.28%).
  • Molecular weight 567.53 g/mol.
  • Chemical formula C26H32Cl2N4O4S.
  • Supplied as a 25 MG package.
  • Appearance: solid; storage: 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.