Learn More
Medchemexpress LLC BMS-199264 hydrochloride | 186180-83-6 | 99.3% | 567.53 g/mol | C26H32Cl2N4O4S | 50 MG
SDP
Catalog No. 5000307930
Encompass_Preferred
BMS-199264 hydrochloride is a research-grade small molecule that inhibits F1F0 ATP hydrolase (IC50 = 0.5 μM) and is supplied as a white to off-white solid for laboratory studies of mitochondrial energetics and ischemia-related ATP decline.
  • High purity: 99.3% (reported by supplier).
  • Chemical formula C26H32Cl2N4O4S and molecular weight 567.53 g/mol.
  • Supplied as milligram-scale packs suitable for bench research.
  • Stable as a solid when stored sealed at 4°C; recommended solvent storage at -20°C to -80°C for extended periods.
  • Provides a defined SMILES/InChI structure for cheminformatics and compound verification.
Catalog No. 50-003-07930 Supplier Medchemexpress LLC Supplier No. HY11896050MG
BMS-199264 hydrochloride is a research-grade small molecule that inhibits F1F0 ATP hydrolase (IC50 = 0.5 μM) and is supplied as a white to off-white solid for laboratory studies of mitochondrial energetics and ischemia-related ATP decline.
  • High purity: 99.3% (reported by supplier).
  • Chemical formula C26H32Cl2N4O4S and molecular weight 567.53 g/mol.
  • Supplied as milligram-scale packs suitable for bench research.
  • Stable as a solid when stored sealed at 4°C; recommended solvent storage at -20°C to -80°C for extended periods.
  • Provides a defined SMILES/InChI structure for cheminformatics and compound verification.