Learn More
Medchemexpress LLC BMS-199264 hydrochloride | 186180-83-6 | MFCD00087197 | 99.3% | 567.53 g·mol⁻¹ | C26H32Cl2N4O4S | 5 MG
SDP
Catalog No. 5000229866
Encompass_Preferred
BMS-199264 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of mitochondrial F1F0 ATP hydrolase. It selectively inhibits ATP hydrolysis (reported IC50 = 0.5 μM), reduces ATP decline during ischemia, and has been reported to improve recovery of contractile function after reperfusion.
  • Inhibits mitochondrial F1F0 ATP hydrolase (IC50 ≈ 0.5 μM).
  • Reduces ATP decline during ischemic conditions.
  • Enhances recovery of contractile function after reperfusion.
  • Provided as a hydrochloride salt for improved stability.
  • High purity suitable for research applications (~99.3%).
  • Available in small research quantities, e.g., 5 MG.
Catalog No. 50-002-29866 Supplier Medchemexpress LLC Supplier No. HY1189605MG
BMS-199264 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of mitochondrial F1F0 ATP hydrolase. It selectively inhibits ATP hydrolysis (reported IC50 = 0.5 μM), reduces ATP decline during ischemia, and has been reported to improve recovery of contractile function after reperfusion.
  • Inhibits mitochondrial F1F0 ATP hydrolase (IC50 ≈ 0.5 μM).
  • Reduces ATP decline during ischemic conditions.
  • Enhances recovery of contractile function after reperfusion.
  • Provided as a hydrochloride salt for improved stability.
  • High purity suitable for research applications (~99.3%).
  • Available in small research quantities, e.g., 5 MG.