Learn More
Medchemexpress LLC HY-19745 10mg Medchemexpress, BMS-202 CAS:1675203-84-5 Purity:>98%
SDP
Catalog No. 502029292
Encompass_Preferred
Medchemexpress, HY-19745 10mg BMS-202 CAS:1675203-84-5 BMS-202 is a potent and nonpeptidic PD-1/PD-L1 complex inhibitor with an IC50 of 18 nM and a KD of 8 μM. BMS-202 binds to PD-L1 and blocks human PD-1/PD-L1 interaction. BMS-202 has antitumor activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9292 Supplier Medchemexpress LLC Supplier No. HY1974510MG
Medchemexpress, HY-19745 10mg BMS-202 CAS:1675203-84-5 BMS-202 is a potent and nonpeptidic PD-1/PD-L1 complex inhibitor with an IC50 of 18 nM and a KD of 8 μM. BMS-202 binds to PD-L1 and blocks human PD-1/PD-L1 interaction. BMS-202 has antitumor activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.