Learn More
Medchemexpress LLC BMS-214662 (hydrochloride) | 195981-08-9 | 98.5% | 526.1 | 25 MG
SDP
Catalog No. 5000394322
Encompass_Preferred
BMS-214662 hydrochloride is a laboratory chemical that acts as an inhibitor. It is provided as a solid and is stable under recommended storage conditions, suitable for various research applications.
  • Stable under recommended storage conditions
  • Store at -20°C
  • Protect from light
  • Store under nitrogen
  • Used for laboratory research
Catalog No. 50-003-94322 Supplier Medchemexpress LLC Supplier No. 25MGHY16111A
BMS-214662 hydrochloride is a laboratory chemical that acts as an inhibitor. It is provided as a solid and is stable under recommended storage conditions, suitable for various research applications.
  • Stable under recommended storage conditions
  • Store at -20°C
  • Protect from light
  • Store under nitrogen
  • Used for laboratory research