Learn More
Medchemexpress LLC BMS 299897 | 290315-45-6 | 99.6% | C24H21ClF3NO4S | 5 MG
SDP
Catalog No. 5000359020
Encompass_Preferred
BMS 299897 is a sulfonamide γ-secretase inhibitor that inhibits Aβ production with an IC50 of 7 nM in HEK293 cells overexpressing amyloid precursor protein (APP).
  • Reduces levels of Aβ peptides.
  • Decreases Aβ peptides to 20-50% of vehicle control at 1 μM.
  • Reduces retrograde QD-BDNF signals, increases anterograde signals.
  • Shows dose- and time-dependent reductions of Aβ in brain, CSF, and plasma in mice.
  • Reduces brain and plasma Aβ1-40.
  • Increases brain concentrations of APP-carboxy-terminal fragments.
  • Attenuates Aβ25-35-induced Aβ1-42 seeding and toxicity.
  • Blocks Aβ25-35-induced deficits in spontaneous alternation or novel object recognition.
Catalog No. 50-003-59020 Supplier Medchemexpress LLC Supplier No. HY508835MG
BMS 299897 is a sulfonamide γ-secretase inhibitor that inhibits Aβ production with an IC50 of 7 nM in HEK293 cells overexpressing amyloid precursor protein (APP).
  • Reduces levels of Aβ peptides.
  • Decreases Aβ peptides to 20-50% of vehicle control at 1 μM.
  • Reduces retrograde QD-BDNF signals, increases anterograde signals.
  • Shows dose- and time-dependent reductions of Aβ in brain, CSF, and plasma in mice.
  • Reduces brain and plasma Aβ1-40.
  • Increases brain concentrations of APP-carboxy-terminal fragments.
  • Attenuates Aβ25-35-induced Aβ1-42 seeding and toxicity.
  • Blocks Aβ25-35-induced deficits in spontaneous alternation or novel object recognition.