Learn More
Medchemexpress LLC HY-18304 5mg Medchemexpress, BMS-3 CAS:1338247-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY183045MG
Encompass_Preferred
Medchemexpress, HY-18304 5mg BMS-3 CAS:1338247-30-5 BMS-3 is a potent LIMK inhibitor with IC50s of 5 nM and 6 nM for LIMK1 and LIMK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2034