Learn More
TARGETMOL CHEMICALS INC BMS-309403 25MG
SDP
Catalog No. 502579872
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-309403 is an effective selective and cell-permeable inhibitor of fatty acid binding protein 4 (FABP4) with a Ki of less than 2 nM. purity: 98%
Catalog No. 50-257-9872 Supplier TARGETMOL CHEMICALS INC Supplier No. T453425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-309403 is an effective selective and cell-permeable inhibitor of fatty acid binding protein 4 (FABP4) with a Ki of less than 2 nM. purity: 98%