Learn More
AdipoGen BMS-309403 (25 mg)
SDP
Supplier:  AdipoGen AGCR13640M025
Encompass_Preferred
Chemical. CAS: 300657-03-8. Formula: C31H26N2O3. MW: 474.6. Cell permeable, potent and selective fatty acid binding protein 4 (FABP4; A-FABP; ALBP; adipocyteP2 protein) inhibitor that competitively targets the fatty acid-binding pocket (Ki= <2nM). Inhibits FABP3 (muscle) and FABP5 (epidermal) with lower affinity (Ki=250nM and 350nM, respectively). FABP4 is an intracellular lipid-binding protein responsible for the transportation of fatty acids. It is expressed primarily in adipose tissue and is associated with inflammation, obesity, diabetes and cardiovascular diseases. Glucose uptake stimulator. Reduces blood glucose levels and increases insulin sensitivity in a mouse model of obesity. Protects against the development of insulin resistance associated with genetic or diet-induced obesity in mice. Anti-atherosclerotic. Decreases fatty acid uptake in adipocytes in vitro and reduces atherosclerotic lesion area in a mouse model of atherosclerosis.
Catalog No. 50-205-4015