Learn More
Apexbio Technology LLC BMS 309403, 5mg. Cas: 300657-03-8 MFCD: MFCD09991687. FABP4 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B77945
Encompass_Preferred
BMS309403 is specifically designed to target FABP4 with a Ki value less than 2 nM. It is a potent and selective FABP4 inhibitor. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5373