Learn More
Medchemexpress LLC BMS-309403 sodium | 2802523-05-1 | 99.9% | 496.53 g/mol | C31H25N2NaO3 | 10 MG
SDP
Catalog No. 5000246806
Encompass_Preferred
BMS-309403 sodium is the sodium salt form of a potent, selective adipocyte fatty acid-binding protein 4 (FABP4) inhibitor used as a research reagent in metabolism and inflammation studies. It is supplied as a high-purity solid in small quantities, intended for analytical, in vitro, and in vivo research applications.
  • Potent and selective FABP4 inhibition for mechanistic studies.
  • High purity suitable for analytical and research use.
  • Available in multiple small solid amounts for dose-finding.
  • Molecular weight 496.53 g/mol for formula-based calculations.
  • Provided with datasheet and safety documentation for reproducibility.
Catalog No. 50-002-46806 Supplier Medchemexpress LLC Supplier No. HY101903A10MG
BMS-309403 sodium is the sodium salt form of a potent, selective adipocyte fatty acid-binding protein 4 (FABP4) inhibitor used as a research reagent in metabolism and inflammation studies. It is supplied as a high-purity solid in small quantities, intended for analytical, in vitro, and in vivo research applications.
  • Potent and selective FABP4 inhibition for mechanistic studies.
  • High purity suitable for analytical and research use.
  • Available in multiple small solid amounts for dose-finding.
  • Molecular weight 496.53 g/mol for formula-based calculations.
  • Provided with datasheet and safety documentation for reproducibility.