Learn More
Medchemexpress LLC BMS-309403 sodium | 2802523-05-1 | MFCD09991687 | 99.9% | 496.53 | C31H25N2NaO3 | 100 MG
SDP
Catalog No. 5000249155
Encompass_Preferred
BMS-309403 sodium is the sodium salt form of a potent, orally active, and selective adipocyte fatty acid binding protein 4 (FABP4) inhibitor used in research on lipid metabolism, metabolic and cardiovascular disease, and endothelial function. The compound is supplied as a solid with batch-specific analytical data available to support research use.
  • High purity suitable for research applications.
  • Selective FABP4 inhibition (Ki <2 nM).
  • Sodium salt form to aid handling and formulation.
  • Batch certificate of analysis available for verification.
  • Documented activity in cellular and animal models.
Catalog No. 50-002-49155 Supplier Medchemexpress LLC Supplier No. HY101903A100MG
BMS-309403 sodium is the sodium salt form of a potent, orally active, and selective adipocyte fatty acid binding protein 4 (FABP4) inhibitor used in research on lipid metabolism, metabolic and cardiovascular disease, and endothelial function. The compound is supplied as a solid with batch-specific analytical data available to support research use.
  • High purity suitable for research applications.
  • Selective FABP4 inhibition (Ki <2 nM).
  • Sodium salt form to aid handling and formulation.
  • Batch certificate of analysis available for verification.
  • Documented activity in cellular and animal models.