Learn More
Medchemexpress LLC HY-10518 50mg Medchemexpress, BMS-345541 BMS-345541 hydrochloride CAS:547757-23-3 Purity:98%
SDP
Catalog No. 501637272
Encompass_Preferred
Medchemexpress,HY-10518 50mg BMS-345541 BMS-345541 hydrochloride CAS:547757-23-3 Formula:C14H18ClN5 BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7272 Supplier Medchemexpress LLC Supplier No. HY1051850MG
Medchemexpress,HY-10518 50mg BMS-345541 BMS-345541 hydrochloride CAS:547757-23-3 Formula:C14H18ClN5 BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.