Learn More
Medchemexpress LLC HY-10519 5mg Medchemexpress, BMS-345541 CAS:445430-58-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105195MG
Encompass_Preferred
Medchemexpress, HY-10519 5mg BMS-345541 CAS:445430-58-0 BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8258