Learn More
Medchemexpress LLC HY-10518 10mg Medchemexpress, BMS-345541 (hydrochloride) CAS:547757-23-3 Purity:>98%
SDP
Catalog No. 502028766
Encompass_Preferred
Medchemexpress, HY-10518 10mg BMS-345541 (hydrochloride) CAS:547757-23-3 BMS-345541 hydrochloride is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8766 Supplier Medchemexpress LLC Supplier No. HY1051810MG
Medchemexpress, HY-10518 10mg BMS-345541 (hydrochloride) CAS:547757-23-3 BMS-345541 hydrochloride is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.