Learn More
Medchemexpress LLC BMS-345541 hydrochloride | 547757-23-3 | 99.9% | 291.78 g·mol⁻¹ | C14H18ClN5 | 10 MM 1 ML
SDP
Catalog No. 5000244097
Encompass_Preferred
BMS-345541 hydrochloride is a selective small-molecule inhibitor of the catalytic subunits of IκB kinase (IKK). It binds at an allosteric site and preferentially inhibits IKK-2 (IC50 0.3 μM) over IKK-1 (IC50 4 μM). The compound is supplied for research use and is available as a solid or as a 10 mM solution in DMSO.
  • Selective inhibition of IκB kinase catalytic subunits.
  • Allosteric binding mechanism provides distinct mode of action.
  • Potent IKK-2 inhibition (IC50 0.3 μM).
  • High reported purity (≈99.9%).
  • Available as solution in DMSO and multiple solid sizes for experimental flexibility.
  • Intended for research use only.
Catalog No. 50-002-44097 Supplier Medchemexpress LLC Supplier No. HY1051810MM 1ML
BMS-345541 hydrochloride is a selective small-molecule inhibitor of the catalytic subunits of IκB kinase (IKK). It binds at an allosteric site and preferentially inhibits IKK-2 (IC50 0.3 μM) over IKK-1 (IC50 4 μM). The compound is supplied for research use and is available as a solid or as a 10 mM solution in DMSO.
  • Selective inhibition of IκB kinase catalytic subunits.
  • Allosteric binding mechanism provides distinct mode of action.
  • Potent IKK-2 inhibition (IC50 0.3 μM).
  • High reported purity (≈99.9%).
  • Available as solution in DMSO and multiple solid sizes for experimental flexibility.
  • Intended for research use only.