Learn More
Medchemexpress LLC BMS-37 | 1675202-20-6 | C27H32N2O4 | 50 MG
SDP
Catalog No. 5000375467
Encompass_Preferred
BMS-37 is a potent PD-1/PD-L1 immune checkpoint inhibitor, demonstrating an IC50 towards the PD-L1/PD-L1 complex within the range of 18 to 200 nM. This compound is valuable for research into PD-L1-induced T-cell exhaustion or as a PD-L1 ligand for the synthesis of PROTAC molecules.
  • Inhibits PD-1/PD-L1 immune checkpoint
  • IC50 towards PD-L1/PD-1 complex between 18 to 200 nM
  • Useful for studying PD-L1-induced T-cell exhaustion
  • Can serve as a PD-L1 ligand for PROTAC molecule synthesis
Catalog No. 50-003-75467 Supplier Medchemexpress LLC Supplier No. HY15087950MG
BMS-37 is a potent PD-1/PD-L1 immune checkpoint inhibitor, demonstrating an IC50 towards the PD-L1/PD-L1 complex within the range of 18 to 200 nM. This compound is valuable for research into PD-L1-induced T-cell exhaustion or as a PD-L1 ligand for the synthesis of PROTAC molecules.
  • Inhibits PD-1/PD-L1 immune checkpoint
  • IC50 towards PD-L1/PD-1 complex between 18 to 200 nM
  • Useful for studying PD-L1-induced T-cell exhaustion
  • Can serve as a PD-L1 ligand for PROTAC molecule synthesis