Learn More
Selleck Chemical LLC BMS-378806 S2632-5mg
SDP
Catalog No. 503110223
Encompass_Preferred
BMS-378806 (BMS 806) selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0 85-26 5 nM in virus
Catalog No. 50-311-0223 Supplier Selleck Chemical LLC Supplier No. S26325MG
BMS-378806 (BMS 806) selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0 85-26 5 nM in virus