Learn More
Medchemexpress LLC HY-14134 5mg Medchemexpress, BMS-378806 CAS:357263-13-9 Purity:>98%
SDP
Catalog No. 502081564
Encompass_Preferred
Medchemexpress, HY-14134 5mg BMS-378806 CAS:357263-13-9 BMS-378806 is a potent HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions. BMS-378806 selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0.85-26.5 nM in virus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1564 Supplier Medchemexpress LLC Supplier No. HY141345MG
Medchemexpress, HY-14134 5mg BMS-378806 CAS:357263-13-9 BMS-378806 is a potent HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions. BMS-378806 selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0.85-26.5 nM in virus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.