Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-433771 dihydrochloride hydrate | 543700-67-0 | 98.0% | 1 ML
SDP

Catalog No. 5000232067
Encompass_Preferred

This dihydrochloride hydrate is a respiratory syncytial virus (RSV) fusion inhibitor supplied as a 10 mM solution in DMSO for research use. It is provided with supporting analytical data and handling guidance.

  • Orally active RSV fusion inhibitor.
  • Supplied as 10 mM solution in DMSO, 1 ML.
  • Purity 98.0%.
  • Molecular weight 468.38 g/mol.
  • Appearance off-white to light yellow solid (supplied as solution).
  • Storage in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-32067 Supplier Medchemexpress LLC Supplier No. HY120632A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This dihydrochloride hydrate is a respiratory syncytial virus (RSV) fusion inhibitor supplied as a 10 mM solution in DMSO for research use. It is provided with supporting analytical data and handling guidance.

  • Orally active RSV fusion inhibitor.
  • Supplied as 10 mM solution in DMSO, 1 ML.
  • Purity 98.0%.
  • Molecular weight 468.38 g/mol.
  • Appearance off-white to light yellow solid (supplied as solution).
  • Storage in solvent: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.