Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS 466442 | 1598424-76-0 | 99.0% | 100 MG
SDP

Catalog No. 5000233917
Encompass_Preferred

BMS-466442 is a potent, selective inhibitor of the Asc-1 (alanine serine cysteine transporter-1) used in neuroscience research and in vitro transporter assays. It is supplied as a high-purity powder with characterized solubility and storage recommendations.

  • Potent asc-1 inhibitor with reported IC50 ~11 nM
  • Inhibits [3H] D-serine uptake in rat synaptosomes (IC50 ~400 nM)
  • Purity reported as 98.99%
  • Molecular weight 538.59 g/mol; formula C31H30N4O5
  • Soluble in DMSO at 100 mg/mL (185.67 mM)
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-33917 Supplier Medchemexpress LLC Supplier No. HY120170100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-466442 is a potent, selective inhibitor of the Asc-1 (alanine serine cysteine transporter-1) used in neuroscience research and in vitro transporter assays. It is supplied as a high-purity powder with characterized solubility and storage recommendations.

  • Potent asc-1 inhibitor with reported IC50 ~11 nM
  • Inhibits [3H] D-serine uptake in rat synaptosomes (IC50 ~400 nM)
  • Purity reported as 98.99%
  • Molecular weight 538.59 g/mol; formula C31H30N4O5
  • Soluble in DMSO at 100 mg/mL (185.67 mM)
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.