Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bms-496 | 2407854-31-1 | 99.7% | 570.39 | 50 MG
SDP

Catalog No. 5000359464
Encompass_Preferred

BMS-496 is a dual DGKα/ζ lipid kinase inhibitor, exhibiting an IC50 of 0.09 μM for DGKα and 0.006 μM for DGKζ. This product is intended for research use only.

  • Dual DGKα/ζ lipid kinase inhibitor
  • Has IC50 of 0.09 μM for DGKα
  • Has IC50 of 0.006 μM for DGKζ
  • Intended for research use only
  • Molecular weight: 570.39
  • Chemical formula: C26H22BrF2N5O3
  • Appearance: Solid
  • Color: Light yellow to yellow

Catalog No. 50-003-59464 Supplier Medchemexpress LLC Supplier No. HY14966650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-496 is a dual DGKα/ζ lipid kinase inhibitor, exhibiting an IC50 of 0.09 μM for DGKα and 0.006 μM for DGKζ. This product is intended for research use only.

  • Dual DGKα/ζ lipid kinase inhibitor
  • Has IC50 of 0.09 μM for DGKα
  • Has IC50 of 0.006 μM for DGKζ
  • Intended for research use only
  • Molecular weight: 570.39
  • Chemical formula: C26H22BrF2N5O3
  • Appearance: Solid
  • Color: Light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.