Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-5 | 1338247-35-0 | 431.29 | 1 ML
SDP

Catalog No. 5000387692
Encompass_Preferred

BMS-5 (LIMKi 3) is a potent LIM kinase (LIMK) inhibitor with IC50s of 7 nM and 8 nM for LIMK1 and LIMK2, respectively. This compound is used in cancer research and inhibits cofilin-Ser3 phosphorylation in a dose-dependent manner.

  • Potent LIM kinase inhibitor
  • IC50 of 7 nM for LIMK1
  • IC50 of 8 nM for LIMK2
  • Target: LIM Kinase
  • Research area: Cancer
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: 100 mg/mL
  • Reduces Nf2∆Ex2 MSC viability

Catalog No. 50-003-87692 Supplier Medchemexpress LLC Supplier No. HY1830510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-5 (LIMKi 3) is a potent LIM kinase (LIMK) inhibitor with IC50s of 7 nM and 8 nM for LIMK1 and LIMK2, respectively. This compound is used in cancer research and inhibits cofilin-Ser3 phosphorylation in a dose-dependent manner.

  • Potent LIM kinase inhibitor
  • IC50 of 7 nM for LIMK1
  • IC50 of 8 nM for LIMK2
  • Target: LIM Kinase
  • Research area: Cancer
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: 100 mg/mL
  • Reduces Nf2∆Ex2 MSC viability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.