Learn More
Medchemexpress LLC BMS-502 | 2407854-18-4 | 99.3% | 516.50 | 100 MG
SDP
Catalog No. 5000361786
Encompass_Preferred
BMS-502 is a potent dual inhibitor of diacylglycerol kinase (DGK) α and ζ, with IC50 values of 4.6 nM and 2.1 nM, respectively. It has been shown to enhance T cell immune responses in mice and can be utilized in tumor immunity related research.
  • Potent dual inhibitor of diacylglycerol kinase alpha and zeta.
  • Enhances T cell immune responses.
  • Useful in tumor immunity research.
  • Has an EC50 of 340 nM in mouse cytotoxic T cell IFN-gamma assay.
  • Demonstrates dose-dependent immune stimulation in mouse models.
Catalog No. 50-003-61786 Supplier Medchemexpress LLC Supplier No. HY149874100MG
BMS-502 is a potent dual inhibitor of diacylglycerol kinase (DGK) α and ζ, with IC50 values of 4.6 nM and 2.1 nM, respectively. It has been shown to enhance T cell immune responses in mice and can be utilized in tumor immunity related research.
  • Potent dual inhibitor of diacylglycerol kinase alpha and zeta.
  • Enhances T cell immune responses.
  • Useful in tumor immunity research.
  • Has an EC50 of 340 nM in mouse cytotoxic T cell IFN-gamma assay.
  • Demonstrates dose-dependent immune stimulation in mouse models.