Learn More
Selleck Chemical LLC BMS-502-E1896-5MG
SDP
Catalog No. 503137681
Encompass_Preferred
BMS-502 is a potent dual inhibitor of diacylglycerol kinase (DGK) and with IC50 values of 4 6 nM and 2 1 nM respectively BMS-502 also exhibits an EC50 value of 340 nM in the mouse cytotoxic T cell IFN- assay (mCTC) BMS-502 can be used in tumor immunity-related research
Catalog No. 50-313-7681 Supplier Selleck Chemical LLC Supplier No. E18965MG
BMS-502 is a potent dual inhibitor of diacylglycerol kinase (DGK) and with IC50 values of 4 6 nM and 2 1 nM respectively BMS-502 also exhibits an EC50 value of 340 nM in the mouse cytotoxic T cell IFN- assay (mCTC) BMS-502 can be used in tumor immunity-related research