Learn More
Medchemexpress LLC HY-10262 10mg Medchemexpress, BMS-536924 CAS:468740-43-4 Purity:>98%
SDP
Catalog No. 502028716
Encompass_Preferred
Medchemexpress, HY-10262 10mg BMS-536924 CAS:468740-43-4 BMS-536924 is an orally active, competitive and selective insulin-like growth factor receptor (IGF-1R) kinase and insulin receptor (IR) inhibitor with IC50s of 100 nM and 73 nM, respectively. BMS-536924 has anti-cancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8716 Supplier Medchemexpress LLC Supplier No. HY1026210MG
Medchemexpress, HY-10262 10mg BMS-536924 CAS:468740-43-4 BMS-536924 is an orally active, competitive and selective insulin-like growth factor receptor (IGF-1R) kinase and insulin receptor (IR) inhibitor with IC50s of 100 nM and 73 nM, respectively. BMS-536924 has anti-cancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.