Learn More
TARGETMOL CHEMICALS INC BMS-536924 10MG
SDP
Catalog No. 502576520
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-536924 (BMS 536924) is an ATP-competitive IGF-1R/IR inhibitor with IC50 of 100 nM/73 nM modest activity for Mek Fak and Lck with very little activity for Akt1 MAPK1/2. purity: 99%
Catalog No. 50-257-6520 Supplier TARGETMOL CHEMICALS INC Supplier No. T641910MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-536924 (BMS 536924) is an ATP-competitive IGF-1R/IR inhibitor with IC50 of 100 nM/73 nM modest activity for Mek Fak and Lck with very little activity for Akt1 MAPK1/2. purity: 99%