Learn More
Selleck Chemical LLC BMS-536924 5mg 468740-43-4 CS-0117
SDP
Supplier:  Selleck Chemical LLC S10125MG
Encompass_Preferred
BMS-536924 (CS-0117) is an ATP-competitive IGF-1R/IR inhibitor with IC50 of 100 nM/73 nM, modest activity for Mek, Fak, and Lck with very little activity for Akt1, MAPK1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8002