Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-561392 formate | 2922280-85-9 | 98.7% | 522.59 g·mol⁻¹ | C28H34N4O6 | 10 MG
SDP

Catalog No. 5000294835
Encompass_Preferred

BMS-561392 formate is the formate salt of a small-molecule inhibitor of TNF-alpha-converting enzyme (TACE/ADAM17) used for preclinical research. Supplied for research use only, it is provided as a high-purity solid suitable for biochemical and cellular assays investigating inflammatory pathways.

  • Inhibits TACE (ADAM17) activity in biochemical assays.
  • High reported purity of 98.69% suitable for research.
  • Molecular formula C28H34N4O6 and molecular weight 522.59 g·mol⁻¹.
  • Supplied in small-mass packages for laboratory use.
  • Store sealed, away from moisture; recommended cold storage.

Catalog No. 50-002-94835 Supplier Medchemexpress LLC Supplier No. HY19667A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-561392 formate is the formate salt of a small-molecule inhibitor of TNF-alpha-converting enzyme (TACE/ADAM17) used for preclinical research. Supplied for research use only, it is provided as a high-purity solid suitable for biochemical and cellular assays investigating inflammatory pathways.

  • Inhibits TACE (ADAM17) activity in biochemical assays.
  • High reported purity of 98.69% suitable for research.
  • Molecular formula C28H34N4O6 and molecular weight 522.59 g·mol⁻¹.
  • Supplied in small-mass packages for laboratory use.
  • Store sealed, away from moisture; recommended cold storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.