Learn More
TARGETMOL CHEMICALS INC BMS-561392 FORMIC ACID 5MG
SDP
Catalog No. 502580408
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-561392 Formic acid is a selective inhibitor of TACE and reduces TNFalpha levels.
Catalog No. 50-258-0408 Supplier TARGETMOL CHEMICALS INC Supplier No. T30531L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-561392 Formic acid is a selective inhibitor of TACE and reduces TNFalpha levels.